Stampa…

Benzo[b]thiophene-3-carboxylic acid ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 1-Benzothiophene-3-carboxylic acid

B4650-200MG B4650-1G
TCIAB4650-200MGEA 36 EUR
TCIAB4650-200MG TCIAB4650-1G
Benzo[b]thiophene-3-carboxylic acid ≥98.0% (by HPLC, titration analysis)
Benzo[b]thiophene-3-carboxylic acid
Formula: C₉H₆O₂S
Peso molecolare: 178,22 g/mol
MDL Number: MFCD01846406
CAS Number: 5381-25-9

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 178.0 to 182.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR