2,4-Bis[8-hydroxy-1,1,7,7-tetramethyljulolidin-9-yl]squaraine ≥98.0% (by total nitrogen basis)
Fornitore: TCI
Synonyms:
2,4-Bis[8-hydroxy-1,1,7,7-tetramethyl-1,2,3,5,6,7-hexahydropyrido[3,2,1-ij]quinoline-9-yl]squaraine
Formula:
C₃₆H₄₄N₂O₄ Peso molecolare: 568.76 g/mol |
MDL Number:
MFCD28386094 CAS Number: 358727-55-6 |
Risultati del test di specificazione
Appearance | Yellow to Amber powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...