6,6'-Bis(chloromethyl)-2,2'-bipyridyl ≥98.0% (by GC, titration analysis)
Fornitore: TCI
|
Pericolo
|
Synonyms:
6,6'-Bis(chloromethyl)-2,2'-bipyridine
Formula:
C₁₂H₁₀Cl₂N₂ |
MDL Number:
MFCD02181082 CAS Number: 74065-64-8 UN: 3261 ADR: 8,III |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 160.0 to 164.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...