3-Butoxyphenylboronic acid (contains varying amounts of Anhydride)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
3-Butoxybenzeneboronic Acid
B4640-1G
B4640-5G
TCIAB4640-1GEA
40
EUR
TCIAB4640-1G
TCIAB4640-5G
3-Butoxyphenylboronic acid (contains varying amounts of Anhydride)
3-Butoxyphenylboronic acid
Formula:
C₁₀H₁₅BO₃ Peso molecolare: 194,04 g/mol |
MDL Number:
MFCD03427053 CAS Number: 352534-81-7 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 97.0 to 110.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...