Stampa…

4-Benzyloxy-3-fluorophenylboronic acid (contains varying amounts of Anhydride)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-Benzyloxy-3-fluorobenzeneboronic acid

B4639-1G B4639-5G
TCIAB4639-1GEA 30 EUR
TCIAB4639-1G TCIAB4639-5G
4-Benzyloxy-3-fluorophenylboronic acid (contains varying amounts of Anhydride)
4-Benzyloxy-3-fluorophenylboronic acid
Formula: C₁₃H₁₂BFO₃
Peso molecolare: 246,04 g/mol
MDL Number: MFCD00994627
CAS Number: 133057-83-7

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 108.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR