5-Bromo-2-chloro-4-methylpyridine ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
5-Bromo-2-chloro-4-picoline
B4626-5G
B4626-25G
TCIAB4626-5GEA
27
EUR
TCIAB4626-5G
TCIAB4626-25G
5-Bromo-2-chloro-4-methylpyridine ≥98.0% (by GC)
5-Bromo-2-chloro-4-methylpyridine
Formula:
C₆H₅BrClN Peso molecolare: 206,47 g/mol |
MDL Number:
MFCD03427663 CAS Number: 778611-64-6 |
Risultati del test di specificazione
Appearance | White or Colorless to Light yellow powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...