Stampa…

1-Boc-piperidine ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: tert-Butyl Piperidine-1-carboxylate, 1-(tert-Butoxycarbonyl)piperidine, Piperidine-1-carboxylic acid tert-Butyl ester, tert-butyl piperidine-1-carboxylate, 1-Boc-piperidine

B4625-25G B4625-5G
TCIAB4625-25GEA 174 EUR
TCIAB4625-25G TCIAB4625-5G
1-Boc-piperidine ≥98.0% (by GC)
1-Boc-piperidine
Formula: C₁₀H₁₉NO₂
Peso molecolare: 185,27 g/mol
Punto di ebollizione: 120°C
Flash Pt: 86°C
MDL Number: MFCD02093935
CAS Number: 75844-69-8

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid to slightly cloudy liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR