Stampa…

(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(tert-butoxy)-4-oxobutanoic acid ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-aspartic Acid 4-tert-Butyl Ester, N-Fmoc-D-aspartic Acid 4-tert-Butyl Ester, 4-tert-Butyl N-Fmoc-D-aspartate, Fmoc-D-Asp(OtBu)-OH

B4619-1G B4619-5G
TCIAB4619-1GEA 20 EUR
TCIAB4619-1G TCIAB4619-5G
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(tert-butoxy)-4-oxobutanoic acid ≥98.0% (by HPLC, titration analysis)
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(tert-butoxy)-4-oxobutanoic acid
Formula: C₂₃H₂₅NO₆
Peso molecolare: 411,5 g/mol
MDL Number: MFCD00077050
CAS Number: 112883-39-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystaline
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D +22.0 to +27.0 deg (C=1,DMF)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR