Stampa…

Benzylthiourea ≥98.0% (by HPLC)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 1-Benzyl-2-thiourea, N-Benzylthiourea

B4612-1G B4612-5G
TCIAB4612-1GEA 29 EUR
TCIAB4612-1G TCIAB4612-5G
Benzylthiourea ≥98.0% (by HPLC)
Benzylthiourea
Formula: C₈H₁₀N₂S
MDL Number: MFCD00041370
CAS Number: 621-83-0
EINECS: 210-709-9
UN: 2811
ADR: 6.1,III

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 162.0 to 166.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR