4,4'-Bis(dodecyloxy)-3-methylazobenzene ≥93.0% (by HPLC)
Fornitore: TCI EUROPE
B4598-500MG
B4598-5G
TCIAB4598-500MGEA
106
EUR
TCIAB4598-500MG
TCIAB4598-5G
4,4'-Bis(dodecyloxy)-3-methylazobenzene ≥93.0% (by HPLC)
4,4'-Bis(dodecyloxy)-3-methylazobenzene
Formula:
C₃₇H₆₀N₂O₂ Peso molecolare: 564,90 g/mol |
CAS Number:
1440509-01-2 |
Risultati del test di specificazione
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (HPLC) | min. 93.0 area% |
Elemental analysis (Nitrogen) | 4.50 to 5.50 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...