Stampa…

4,4'-Bis(decyloxy)-3-methylazobenzene ≥95.0% (by HPLC)

Fornitore: TCI EUROPE

B4597-500MG B4597-5G
TCIAB4597-500MGEA 119 EUR
TCIAB4597-500MG TCIAB4597-5G
4,4'-Bis(decyloxy)-3-methylazobenzene ≥95.0% (by HPLC)
4,4'-Bis(decyloxy)-3-methylazobenzene
Formula: C₃₃H₅₂N₂O₂
Peso molecolare: 508,79 g/mol
CAS Number: 1627844-78-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d8953752b6c68bc001e4ab02de114f9429de6eec&vid=d8953752b6c68bc001e4ab02de114f9429de6eec&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 95.0 area%
Elemental analysis (Nitrogen) 5.20 to 6.20 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR