4,4'-Bis(hexyloxy)-3-methylazobenzene ≥95.0% (by HPLC)
Fornitore: TCI EUROPE
B4596-500MG
B4596-5G
TCIAB4596-500MGEA
73
EUR
TCIAB4596-500MG
TCIAB4596-5G
4,4'-Bis(hexyloxy)-3-methylazobenzene ≥95.0% (by HPLC)
4,4'-Bis(hexyloxy)-3-methylazobenzene
Formula:
C₂₅H₃₆N₂O₂ Peso molecolare: 396,58 g/mol |
CAS Number:
1440509-03-4 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystaline |
Purity (HPLC) | min. 95.0 area% |
Elemental analysis (Nitrogen) | 6.50 to 7.50 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...