2,4-Bis(benzyloxy)-6-chloro-1,3,5-triazine ≥95.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
DBT-Cl
B4587-200MG
B4587-1G
TCIAB4587-200MGEA
47
EUR
TCIAB4587-200MG
TCIAB4587-1G
2,4-Bis(benzyloxy)-6-chloro-1,3,5-triazine ≥95.0% (by HPLC, titration analysis)
2,4-Bis(benzyloxy)-6-chloro-1,3,5-triazine
Formula:
C₁₇H₁₄ClN₃O₂ Peso molecolare: 327,77 g/mol |
CAS Number:
851030-18-7 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Purity (Argentmetric Titration) | min. 95.0 % |
Melting point | 64.0 to 68.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...