7-Bromo-2-naphthol ≥97.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
7-Bromonaphthalen-2-ol, 7-Bromo-2-hydroxynaphthalene, 2-Bromo-7-hydroxynaphthalene
B4580-1G
B4580-5G
TCIAB4580-1GEA
25
EUR
TCIAB4580-1G
TCIAB4580-5G
7-Bromo-2-naphthol ≥97.0% (by GC)
7-Bromo-2-naphthol
Formula:
C₁₀H₇BrO Peso molecolare: 223,07 g/mol |
MDL Number:
MFCD00277644 CAS Number: 116230-30-9 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 132.0 to 136.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...