Stampa…

Butyltin trichloride ≥98.0% (by GC)

Fornitore: TCI EUROPE
Pericolo

Synonyms: n-Butyltin trichloride, Trichlorobutylstannane

B4575-25G B4575-100G
TCIAB4575-25GEA 31 EUR
TCIAB4575-25G TCIAB4575-100G
Butyltin trichloride ≥98.0% (by GC)
Butyltin trichloride
Formula: C₄H₉Cl₃Sn
Peso molecolare: 282,18 g/mol
Punto di ebollizione: 93°C
Punto di fusione: –63°C
Flash Pt: 81°C
MDL Number: MFCD00000515
CAS Number: 1118-46-3
EINECS: 214-263-6
UN: 3265
ADR: 8,II

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (Argentmetric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR