Stampa…

9-(4-Biphenylyl)-10-bromoanthracene ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 9-([1,1'-Biphenyl]-4-yl)-10-bromoanthracene

B4569-1G B4569-5G
TCIAB4569-1GEA 65 EUR
TCIAB4569-1G TCIAB4569-5G
9-(4-Biphenylyl)-10-bromoanthracene ≥98.0% (by GC)
9-(4-Biphenylyl)-10-bromoanthracene
Formula: C₂₆H₁₇Br
Peso molecolare: 409,33 g/mol
MDL Number: MFCD19440858
CAS Number: 400607-05-8

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 251.0 to 255.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR