Benzylamine Hydroiodide, low water content ≥98.0% (by HPLC, total nitrogen)
Fornitore: TCI
|
Attenzione
|
B4566-1G
B4566-5G
TCIAB4566-1GEA
63
EUR
TCIAB4566-1G
TCIAB4566-5G
Benzylamine Hydroiodide, low water content ≥98.0% (by HPLC, total nitrogen)
Benzylamine hydroiodide
Formula:
C₇H₉N |
CAS Number:
45579-91-7 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 176.0 to 180.0 deg-C |
Water | max. 100 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...