Bambuterol hydrochloride ≥96.0% (by HPLC, titration analysis)
Fornitore: TCI
Formula:
C₁₈H₂₉N₃O₅ |
MDL Number:
MFCD03427293 CAS Number: 81732-46-9 Merck Index: 14,00953 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 96.0 area% |
Purity (Nonaqueous Titration) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...