Ethyl tridecanoate ≥97%
Fornitore: Thermo Scientific
Synonyms:
Tridecylic acid ethyl ester
Purity: 97%
CAS No.: 28267-29-0
Molecular Formula: C15H30O2
Formula Weight: 242.41
Flash Point:
Formula:
C₁₅H₃₀O₂ Peso molecolare: 242,4 g/mol Punto di ebollizione: 121…123 °C (2,2 mmHg) Densità: 0,86 Flash Pt: >110 °C (230 °F) Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00008978 CAS Number: 28267-29-0 EINECS: 248-929-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...