Stampa…

2,5-Bis(4-bromophenyl)-1,1-dimethyl-3,4-diphenylsilole ≥95.0% (by GC)

Fornitore: TCI

B4547-1G B4547-5G
TCIAB4547-1GEA 152 EUR
TCIAB4547-1G TCIAB4547-5G
2,5-Bis(4-bromophenyl)-1,1-dimethyl-3,4-diphenylsilole ≥95.0% (by GC)
2,5-Bis(4-bromophenyl)-1,1-dimethyl-3,4-diphenylsilole
Formula: C₃₀H₂₄Br₂Si
CAS Number: 866769-99-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f4c7ecb9da5ebfcca6fb5c6d376309d0468568b&vid=0f4c7ecb9da5ebfcca6fb5c6d376309d0468568b&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Purity (GC) min. 95.0 %
Melting point 222.0 to 226.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR