1,3-Bis(4-chlorophenyl)urea ≥98.0% (by HPLC, total nitrogen)
Fornitore: TCI
|
Attenzione
|
Formula:
C₁₃H₁₀Cl₂N₂O |
MDL Number:
MFCD00018541 CAS Number: 1219-99-4 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...