Stampa…

Bis(4-bromophenylboronic acid)scylloinositol complex dipotassium tetrahydrate ≥98.0% (by HPLC)

Fornitore: TCI EUROPE
Attenzione

B4524-200MG B4524-1G
TCIAB4524-200MGEA 33 EUR
TCIAB4524-200MG TCIAB4524-1G
Bis(4-bromophenylboronic acid)scylloinositol complex dipotassium tetrahydrate ≥98.0% (by HPLC)
Bis(4-bromophenylboronic acid)scylloinositol complex dipotassium tetrahydrate
Formula: C₁₈H₁₄B₂Br₂K₂O₆
Peso molecolare: 585,93 g/mol
CAS Number: 1537876-29-1

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 97.0 % (calcd.on anh.substance)
Water 9.5 to 12.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR