Bis(3-bromophenylboronic acid)scyllo inositol complex dipotassium pentahydrate
Fornitore: TCI EUROPE
|
Attenzione
|
B4523-200MG
B4523-1G
TCIAB4523-200MGEA
33
EUR
TCIAB4523-200MG
TCIAB4523-1G
Bis(3-bromophenylboronic acid)scyllo inositol complex dipotassium pentahydrate
Bis(3-bromophenylboronic acid)scyllo inositol complex dipotassium pentahydrate
Formula:
C₁₈H₁₄B₂Br₂K₂O₆ Peso molecolare: 585,93 g/mol |
CAS Number:
1537876-31-5 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 97.0 % (calcd.on anh.substance) |
Water | 12.0 to 14.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...