Stampa…

2-(4-Bromophenyl)-5-phenyl-1,3,4-oxadiazole ≥97.0% (by HPLC)

Fornitore: TCI EUROPE

B4513-1G B4513-5G
TCIAB4513-1GEA 77 EUR
TCIAB4513-1G TCIAB4513-5G
2-(4-Bromophenyl)-5-phenyl-1,3,4-oxadiazole ≥97.0% (by HPLC)
2-(4-Bromophenyl)-5-phenyl-1,3,4-oxadiazole
Formula: C₁₄H₉BrN₂O
Peso molecolare: 301,14 g/mol
MDL Number: MFCD00050997
CAS Number: 21510-43-0
EINECS: 244-413-6

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Argentmetric Titration) min. 95.0 %
Melting point 169.0 to 173.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR