Stampa…

2-Bromo-6-(2-methyl-1,3-dioxolan-2-yl)pyridine ≥98.0% (by GC)

Fornitore: TCI EUROPE

B4512-1G B4512-5G
TCIAB4512-1GEA 23 EUR
TCIAB4512-1G TCIAB4512-5G
2-Bromo-6-(2-methyl-1,3-dioxolan-2-yl)pyridine ≥98.0% (by GC)
2-Bromo-6-(2-methyl-1,3-dioxolan-2-yl)pyridine
Formula: C₉H₁₀BrNO₂
Peso molecolare: 244,09 g/mol
MDL Number: MFCD12406888
CAS Number: 49669-14-9

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 43.0 to 47.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR