Stampa…

2-(4-Bromophenyl)pyridine ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

B4500-1G B4500-5G
TCIAB4500-1GEA 33 EUR
TCIAB4500-1G TCIAB4500-5G
2-(4-Bromophenyl)pyridine ≥98.0% (by GC, titration analysis)
2-(4-Bromophenyl)pyridine
Formula: C₁₁H₈BrN
MDL Number: MFCD04116231
CAS Number: 63996-36-1

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 61.0 to 65.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR