2-Bromo-4-(trifluoromethyl)phenol ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
3-Bromo-4-hydroxybenzotrifluoride
B4492-1G
B4492-5G
TCIAB4492-1GEA
55
EUR
TCIAB4492-1G
TCIAB4492-5G
2-Bromo-4-(trifluoromethyl)phenol ≥98.0% (by GC, titration analysis)
2-Bromo-4-(trifluoromethyl)phenol
Formula:
C₇H₄BrF₃O Peso molecolare: 241,01 g/mol |
MDL Number:
MFCD06657954 CAS Number: 81107-97-3 |
Risultati del test di specificazione
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...