Stampa…

3-Bromo-4-fluorophenol ≥99.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

B4491-5G B4491-25G
TCIAB4491-5GEA 32 EUR
TCIAB4491-5G TCIAB4491-25G
3-Bromo-4-fluorophenol ≥99.0% (by GC, titration analysis)
3-Bromo-4-fluorophenol
Formula: C₆H₄BrFO
Punto di ebollizione: 78°C
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD03425884
CAS Number: 27407-11-0

Visualizza i codici

Risultati del test di specificazione

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 99.0 %
Purity (Neutralization titration) min. 99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR