Stampa…

5-Bromo-2,3-difluorophenol ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 1-Bromo-3,4-difluoro-5-hydroxybenzene

B4490-5G B4490-25G
TCIAB4490-5GEA 29 EUR
TCIAB4490-5G TCIAB4490-25G
5-Bromo-2,3-difluorophenol ≥98.0% (by GC)
5-Bromo-2,3-difluorophenol
Formula: C₆H₃BrF₂O
Peso molecolare: 208,99 g/mol
MDL Number: MFCD01631350
CAS Number: 186590-26-1

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Green clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR