Stampa…

[(1H-Benzotriazol-1-yl)methyl]triphenylphosphonium chloride ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: [(1H-Benzotriazol-1-yl)methyl]triphenylphosphonium Chloride

B4486-1G B4486-5G
TCIAB4486-1GEA 30 EUR
TCIAB4486-1G TCIAB4486-5G
[(1H-Benzotriazol-1-yl)methyl]triphenylphosphonium chloride ≥98.0% (by HPLC, titration analysis)
[(1H-Benzotriazol-1-yl)methyl]triphenylphosphonium chloride
Formula: C₂₅H₂₁ClN₃P
Peso molecolare: 429,89 g/mol
MDL Number: MFCD00960175
CAS Number: 111198-09-5

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR