2-(4-Bromophenyl)-1-phenylbenzimidazole ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
Synonyms:
2-(4-Bromophenyl)-1-phenyl-1H-benzoimidazole
B4479-1G
B4479-5G
TCIAB4479-1GEA
47
EUR
TCIAB4479-1G
TCIAB4479-5G
2-(4-Bromophenyl)-1-phenylbenzimidazole ≥98.0% (by GC, titration analysis)
2-(4-Bromophenyl)-1-phenylbenzimidazole
Formula:
C₁₉H₁₃BrN₂ Peso molecolare: 349,23 g/mol |
MDL Number:
MFCD09261342 CAS Number: 2620-76-0 |
Risultati del test di specificazione
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 166.0 to 170.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...