Stampa…

Benzofurazan ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 2,1,3-Benzoxadiazole, Benzo[c][1,2,5]oxadiazole

B4473-1G B4473-5G
TCIAB4473-1GEA 60 EUR
TCIAB4473-1G TCIAB4473-5G
Benzofurazan ≥98.0% (by GC)
Benzofurazan
Formula: C₆H₄N₂O
Peso molecolare: 120,11 g/mol
MDL Number: MFCD00068064
CAS Number: 273-09-6

Visualizza i codici

Risultati del test di specificazione

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 51.0 to 55.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR