Stampa…

1-Benzyl-3-methylpiperidin-4-one ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 1-Benzyl-3-methyl-4-piperidone

B4465-1G B4465-5G
TCIAB4465-1GEA 45 EUR
TCIAB4465-1G TCIAB4465-5G
1-Benzyl-3-methylpiperidin-4-one ≥98.0% (by GC, titration analysis)
1-Benzyl-3-methylpiperidin-4-one
Formula: C₁₃H₁₇NO
Peso molecolare: 203,2800 g/mol
Punto di ebollizione: 99°C
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00044806
CAS Number: 34737-89-8
EINECS: 252-176-5
UN: 1760
ADR: 8,III

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR