Stampa…

Beclomethasone dipropionate ≥98.0% (by HPLC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Beclometasone dipropionate

B4464-1G B4464-5G
TCIAB4464-1GEA 55 EUR
TCIAB4464-1G TCIAB4464-5G
Beclomethasone dipropionate ≥98.0% (by HPLC)
Beclomethasone dipropionate
Formula: C₂₈H₃₇ClO₇
Peso molecolare: 521,047 g/mol
MDL Number: MFCD00135613
CAS Number: 5534-09-8
EINECS: 226-886-0
Merck Index: 14,01019

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Specific rotation [a]20/D +88.0 to +94.0 deg (C=1, Dioxane)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR