Stampa…

2-Bromo-3-decylthiophene ≥97.0% (by GC)

Fornitore: TCI EUROPE

B4455-5G B4455-25G
TCIAB4455-5GEA 54 EUR
TCIAB4455-5G TCIAB4455-25G
2-Bromo-3-decylthiophene ≥97.0% (by GC)
2-Bromo-3-decylthiophene
Formula: C₁₄H₂₃BrS
Peso molecolare: 303,30 g/mol
MDL Number: MFCD16619251
CAS Number: 144012-09-9

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR