8-Bromo-7-(2-butyn-1-yl)-3-methylxanthine ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
Synonyms:
8-Bromo-7-(but-2-yn-1-yl)-3-methyl-1H-purine-2,6(3H,7H)-dione, 8-Bromo-7-(2-butyn-1-yl)-3,7-dihydro-3-methyl-1H-purine-2,6-dione
B4454-200MG
B4454-1G
TCIAB4454-200MGEA
41
EUR
TCIAB4454-200MG
TCIAB4454-1G
8-Bromo-7-(2-butyn-1-yl)-3-methylxanthine ≥98.0% (by HPLC, titration analysis)
8-Bromo-7-(2-butyn-1-yl)-3-methylxanthine
Formula:
C₁₀H₉BrN₄O₂ Peso molecolare: 297,11 g/mol |
MDL Number:
MFCD22126091 CAS Number: 666816-98-4 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...