Stampa…

3-(4-Bromophenyl)-9-phenylcarbazole ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3-(4-Bromophenyl)-9-phenyl-9H-carbazole

B4452-1G B4452-5G
TCIAB4452-1GEA 16 EUR
TCIAB4452-1G TCIAB4452-5G
3-(4-Bromophenyl)-9-phenylcarbazole ≥98.0% (by GC)
3-(4-Bromophenyl)-9-phenylcarbazole
Formula: C₂₄H₁₆BrN
Peso molecolare: 398,30 g/mol
MDL Number: MFCD14582939
CAS Number: 1028647-93-9

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 136.0 to 140.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR