Stampa…

(S)-1-Boc-3-hydroxypiperidine ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: (S)-tert-Butyl 3-hydroxypiperidine-1-carboxylate, (3S)-3-Hydroxypiperidine, N-BOC protected

B4446-1G B4446-5G
TCIAB4446-1GEA 16 EUR
TCIAB4446-1G TCIAB4446-5G
(S)-1-Boc-3-hydroxypiperidine ≥98.0% (by GC)
(S)-1-Boc-3-hydroxypiperidine
Formula: C₁₀H₁₉NO₃
Peso molecolare: 201,27 g/mol
MDL Number: MFCD04115306
CAS Number: 143900-44-1

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 52.0 to 56.0 deg-C
Specific rotation [a]20/D +20.0 to +23.0 deg (C=5,MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR