Bis(4-bromophenyl)sulphide ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Bis(4-bromophenyl)sulphane
B4435-1G
B4435-5G
TCIAB4435-1GEA
40
EUR
TCIAB4435-1G
TCIAB4435-5G
Bis(4-bromophenyl)sulphide ≥98.0% (by GC)
Bis(4-bromophenyl)sulphide
Formula:
C₁₂H₈Br₂S Peso molecolare: 344,06 g/mol Punto di ebollizione: 210°C |
MDL Number:
MFCD00623633 CAS Number: 3393-78-0 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 113.0 to 117.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...