Butylamine hydroiodide ≥97.0% (by total nitrogen and titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
B4433-1G
B4433-5G
TCIAB4433-1GEA
70
EUR
TCIAB4433-1G
TCIAB4433-5G
Butylamine hydroiodide ≥97.0% (by total nitrogen and titration analysis)
Butylamine hydroiodide
Formula:
C₄H₁₁N |
CAS Number:
36945-08-1 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (with Total Nitrogen) | min. 97.0 % |
Purity (Argentmetric Titration) | min. 97.0 % |
Melting point | 169.0 to 173.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...