Stampa…

9-(3-Bromophenyl)carbazole ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 9-(3-Bromophenyl)-9H-carbazole

B4427-1G B4427-5G
TCIAB4427-1GEA 56 EUR
TCIAB4427-1G TCIAB4427-5G
9-(3-Bromophenyl)carbazole ≥98.0% (by GC)
9-(3-Bromophenyl)carbazole
Formula: C₁₈H₁₂BrN
Peso molecolare: 322,21 g/mol
MDL Number: MFCD20486475
CAS Number: 185112-61-2

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 67.0 to 71.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR