2-(4-Biphenylyl)amino-9,9-dimethylfluorene ≥98.0% (by HPLC, total nitrogen)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
N-([1,1'-Biphenyl]-4-yl)-9,9-dimethyl-9H-fluoren-2-amine
B4424-1G
B4424-5G
TCIAB4424-1GEA
29
EUR
TCIAB4424-1G
TCIAB4424-5G
2-(4-Biphenylyl)amino-9,9-dimethylfluorene ≥98.0% (by HPLC, total nitrogen)
2-(4-Biphenylyl)amino-9,9-dimethylfluorene
Formula:
C₂₇H₂₃N Peso molecolare: 361,49 g/mol |
MDL Number:
MFCD16036286 CAS Number: 897671-69-1 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 145.0 to 149.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...