Stampa…

2-Bromo-5-methoxybenzyl bromide ≥98.0% (by GC)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 4-Bromo-3-(bromomethyl)anisole, 1-Bromo-2-(bromomethyl)-4-methoxybenzene

B4423-5G B4423-25G
TCIAB4423-5GEA 48 EUR
TCIAB4423-5G TCIAB4423-25G
2-Bromo-5-methoxybenzyl bromide ≥98.0% (by GC)
2-Bromo-5-methoxybenzyl bromide
Formula: C₈H₈Br₂O
Peso molecolare: 279,96 g/mol
MDL Number: MFCD00078664
CAS Number: 19614-12-1
UN: 3261
ADR: 8,II

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystaline
Purity (GC) min. 98.0 %
Melting point 91.0 to 95.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR