Tricloruro di boro ∼13% ∼1.0 mol/L in toluene
Fornitore: TCI
|
Pericolo
|
Formula:
BCl₃ Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00011313 CAS Number: 10294-34-5 EINECS: 233-658-4 UN: 2924 ADR: 3,II Merck Index: 14,01348 |
Appearance | Colorless to Light orange to Yellow clear liquid |
Concentration (Neutralization titration) | 12.5% to 14.0 w/w% |