Stampa…

Butylmalonic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Butyl-1,3-propanedioic acid, 2-Butylmalonic acid

B4414-5G B4414-25G
TCIAB4414-5GEA 54 EUR
TCIAB4414-5G TCIAB4414-25G
Butylmalonic acid ≥98.0% (by GC, titration analysis)
Butylmalonic acid
Formula: C₇H₁₂O₄
Peso molecolare: 160,17 g/mol
MDL Number: MFCD00002673
CAS Number: 534-59-8
EINECS: 208-602-7
Merck Index: 14,01576

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 101.0 to 105.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR