2-Bromo-5-nitrothiazole ≥97.0% (by GC)
Fornitore: TCI
|
Attenzione
|
Formula:
C₃HBrN₂O₂S |
MDL Number:
MFCD00005317 CAS Number: 3034-48-8 EINECS: 221-226-8 |
Risultati del test di specificazione
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 85.0 to 91.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...