Stampa…

Ammonium ((1S,3R,4R,7S)-3-bromo-1,7-dimethyl-2-oxobicyclo[2.2.1]heptan-7-yl)methanesulphonate ≥97.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: (-)-3-Bromocamphor-8-sulphonic acid Ammonium salt

B4412-1G B4412-5G
TCIAB4412-1GEA 67 EUR
TCIAB4412-1G TCIAB4412-5G
Ammonium ((1S,3R,4R,7S)-3-bromo-1,7-dimethyl-2-oxobicyclo[2.2.1]heptan-7-yl)methanesulphonate ≥97.0% (by titrimetric analysis)
Ammonium ((1S,3R,4R,7S)-3-bromo-1,7-dimethyl-2-oxobicyclo[2.2.1]heptan-7-yl)methanesulphonate
Formula: C₁₀H₁₈BrNO₄S
Peso molecolare: 328,22 g/mol
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD00151070
CAS Number: 55870-50-3
EINECS: 259-871-2

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (Ion exchange titration) min. 97.0 %
Specific rotation [a]20/D -83.0 to -86.0 deg (C=1, H2O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR