Stampa…

4,8-Bis(2-ethylhexyloxy)benzo[1,2-b:4,5-b']dithiophene ≥95.0% (by HPLC)

Fornitore: TCI EUROPE

Synonyms: 4,8-Bis((2-ethylhexyl)oxy)benzo[1,2-b:4,5-b']dithiophene

B4395-200MG B4395-1G B4395-5G
TCIAB4395-200MGEA 50 EUR
TCIAB4395-200MG TCIAB4395-1G TCIAB4395-5G
4,8-Bis(2-ethylhexyloxy)benzo[1,2-b:4,5-b']dithiophene ≥95.0% (by HPLC)
4,8-Bis(2-ethylhexyloxy)benzo[1,2-b:4,5-b']dithiophene
Formula: C₂₆H₃₈O₂S₂
Peso molecolare: 446,71 g/mol
MDL Number: MFCD17018551
CAS Number: 1160823-77-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2d7463624836844e18daac243593b9eeec046df&vid=f2d7463624836844e18daac243593b9eeec046df&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown clear liquid
Purity (HPLC) min. 95.0 area%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR