1-(1-Boc-piperidino)-4-bromo-1H-pyrazole ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
tert-Butyl 4-(4-bromo-1H-pyrazol-1-yl)piperidine-1-carboxylate, 1-(4-Boc-1-piperidinyl)-4-bromopyrazole
B4391-1G
B4391-5G
TCIAB4391-1GEA
38
EUR
TCIAB4391-1G
TCIAB4391-5G
1-(1-Boc-piperidino)-4-bromo-1H-pyrazole ≥98.0% (by HPLC, titration analysis)
1-(1-Boc-piperidino)-4-bromo-1H-pyrazole
Formula:
C₁₃H₂₀BrN₃O₂ Peso molecolare: 330,23 g/mol |
MDL Number:
MFCD09831980 CAS Number: 877399-50-3 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Argentmetric Titration) | min. 98.0 % |
Melting point | 77.0 to 81.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...