Stampa…

2-Bromocarbazole ≥98.0% (by GC)

Fornitore: TCI
Attenzione

B4381-5G B4381-25G
TCIAB4381-5GEA 103 EUR
TCIAB4381-5G TCIAB4381-25G
2-Bromocarbazole ≥98.0% (by GC)
2-Bromocarbazole
Formula: C₁₂H₈BrN
MDL Number: MFCD09750430
CAS Number: 3652-90-2

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 253.0 to 257.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR